Principaux documents
BAF185120000
SP Bel-Art® Scintillation Vial Rack
to hold, holds 24 x 25-30mm Vials
Synonyme(s) :
autoclavable polypropylene
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Caractéristiques
to hold holds 24 x 25-30mm Vials
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18512-0000
H × L × l
3.75 in. (95 mm) × 11.75 in. (298 mm) × 5 in. (127 mm)
Description générale
This rugged and stable rack holds 24 scintillation vials (30mm diameter) for convenient storage, handling, and sample preparation.
Caractéristiques et avantages
  • This rugged and stable rack holds 24 scintillation vials (30mm diameter) for convenient storage, handling, and sample preparation.
  • Corrosion-resistant, autoclavable polypropylene
  • Shelf height: 4.4cm (1³/₄")
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP