Key Documents
Z376825
Sigma-Aldrich
Scintillation vials, general purpose
polypropylene
Synonym(s):
Scintillation Vials
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
Pricing and availability is not currently available.
material
polypropylene
sterility
non-sterile
packaging
pkg of 1000 ea
manufacturer/tradename
Simport S207-5
vial size
16 mm × 57 mm
volume
6.5 mL
General description
General purpose vials can be used for liquid scintillation counting, gamma counting, chromatography, sample storage, and culturing. Fits LKB and Packard Varisette counters. Vials are securely sealed by snapping the cap on. Cap removal requires a simple 1/4 twist, reducing the chance of spillage associated with pull of caps. Polyethylene vials are supplied with polypropylene caps. Polypropylene vials are supplied with polyethylene caps.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)