Key Documents
SYNP160001
Synthware cylindrical pressure vessel, heavy wall
15 mL, O.D. × L 26 mm × 109 mm, #15 bushing
Synonym(s):
Heavy-Wall Pressure Vessel, Synthware P160001, pressure tube
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Synthware P160001
O.D. × L
26 mm × 109 mm
capacity
15 mL
fitting
#15 bushing
Related Categories
General description
Heavy wall cylindrical vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal. This unique design allows hand tightening and loosening for easy access into the vessel. The O-ring is positioned on the bushing above the thread for improved sealing. The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime. Complete apparatus is supplied with vessel, PTFE bushing and Viton® O-ring. *Important Warnings for Pressurized Glassware: Due to varying conditions, we cannot guarantee glass vessels from breakage under pressure. Always work behind a safety shield.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. what is the pressure rating of these bottles
    1 answer
    1. The pressure rating for this item is 88psi.
      Helpful?
  2. What size of Viton O-ring is suitable for Synthware™ cylindrical pressure vessel, heavy wall, 15 ml (product number SYNP160001-1EA)?
    1 answer
    1. Unfortunately, replacement O-rings for this product are not available at this time. These items are available directly from the manufacturer under Synthware product number R111EP. Please see the link below to the supplier product page:
      https://kemtech-america.com/Product/Product_Info.aspx?ProductID=3033
      Helpful?