Skip to Content
Merck
All Photos(1)

Key Documents

SML1013

Sigma-Aldrich

PRT4165

≥98% (HPLC)

Synonym(s):

2-(3-Pyridinylmethylene)-1H-indene-1,3(2H)-dione; 2-(3-Pyridylmethylene)-1,3-indandione, NSC 600157

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9NO2
CAS Number:
Molecular Weight:
235.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C(C(C2=C1C=CC=C2)=O)=CC3=CN=CC=C3

InChI

1S/C15H9NO2/c17-14-11-5-1-2-6-12(11)15(18)13(14)8-10-4-3-7-16-9-10/h1-9H

InChI key

OMHZFEWYVFWVLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

PRT4165 is a potent inhibitor of PRC1 (polycomb-repressive complex 1)-mediated histone H2A ubiquitylation in vivo and in vitro. PRT4165 inhibits the in vitro histone H2A E3 ubiquitin ligase activity of PRC1 components RING1, RNF2, and a BMI1/RNF2 complex. PRT4165 also inhibits DNA double-strand breaks induced ubiquitylation of histone H2AX.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dario Tommasini et al.
iScience, 26(4), 106358-106358 (2023-03-31)
Neurological disease is characterized the by dysfunction of specific neuroanatomical regions. To determine whether region-specific vulnerabilities have a transcriptional basis at cell-type-specific resolution, we analyzed gene expression in mouse oligodendrocytes across various brain regions. Oligodendrocyte transcriptomes cluster in an anatomical

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service