Skip to Content
Merck
All Photos(1)

Key Documents

THP 1 Cell Line human

88081201, human blood, Monocyte

Synonym(s):

O-THP-1 Cells, THP-1 Macrophages, THP-1 Monocytes, THP1 Cells, THPI Cells, Tohoku Hospital Pediatrics-1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

THP 1 Cell Line human, 88081201, Leukemic monocyte

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Monocyte

products

Not specified

receptors

Fc, C3b

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human monocytic leukaemia

Cell Line Description

Derived from the peripheral blood of a 1 year old male with acute monocytic leukaemia. THP-1 cells have Fc and C3b receptors and lack surface and cytoplasmic immunoglobulins. These cells also stain positive for α-napthhyl butyrate esterase, produce lysozymes and are phagocytic (both latex beads and sensitised erythrocytes). THP-1 cells can also restore the response of purified T lymphocytes to Concanavlin A, show increased CO2 production on phagocytosis and can be differentiated into macrophage-like cells using for example DMSO.

Application

THP 1 cell line human has been used to study:
  • bactericidal activity of moxifloxacin against Staphylococcus aureus
  • the effect of lipopolysaccharide-mediated inflammatory response in murine versus human cell lines
  • the effects of purine nucleosides on the in vitro growth of Cryptosporidium parvum
  • the change in gene expression after infection with Mycobacterium tuberculosis
  • the induction of cytokines in the cell line in presence of antigens obtained from Clostridium difficile

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,13
D13S317: 13
D16S539: 11,12
D5S818: 11,12
D7S820: 10
THO1: 8,9.3
TPOX: 8,11
vWA: 16

Subculture Routine

If starting from a frozen ampoule the cryoprotectant should be removed. Add thawed cells to a conical based centrifuge tube, e.g. 15ml tube, slowly add 4 ml of culture medium to the tube. Take a sample of the cell suspension, e.g. 100μl, to count cells. Centrifuge the cell suspension at low speed i.e. 100 - 150 x g for a maximum of 5 minutes. Remove medium and resuspend the cell pellet at a density of 3 - 5 x 100,000 cells/ml in fresh medium containing 20% serum. Incubate flask at 37°C; 5 - 7% CO2. Check daily. Keep flask in a vertical position until the cells reach the exponential phase of growth. This can take up to 7 days. Once the culture is established the serum concentration can be reduced to 10%. To keep the cells in exponential growth, maintain cultures between 3-8x100,000 cells/ml. Requires 5% DMSO and 95% fetal bovine serum (FBS) as cryoprotectant. Growing orders are recommended due to difficulties that can be experienced during the initial start-up of this cell line. Replacements will be charged at full cost where claims cannot be substantiated.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service