Skip to Content
Merck
All Photos(1)

Documents

KG-1

86111306, human bone marrow, Myeloblast-like

Synonym(s):

KG1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

KG-1, 86111306

biological source

human bone marrow

growth mode

Suspension

karyotype

2n = 46, diploid

morphology

Myeloblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian bone marrow myelogenous leukaemia

Cell Line Description

Derived from the bone marrow aspirate of a 59 year old male Caucasian with erythroleukemia that became acute myelogenous leukaemia. The cells form colonies in soft-agar in the presence of colony stimulating factor (CSF). KG-1 cells resemble acute myelogenous leukaemia showing considerable pleomorphism with a predominance of myeloblasts and promyelocytes. A small percentage of the cells are mature granulocytes with the occasional presence of macrophages and eosinophils. They do not express surface Ig or EBV antigens and lack specific markers for lymphocytic cells.

Application

KG-1 has been used to extract total RNA and use them as templates to determine the 5′ end of promyelocytic leukemia zinc finger (PLZF) transcripts.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 7
D13S317: 11,12
D16S539: 10,11
D5S818: 13
D7S820: 8,10
THO1: 7,8
TPOX: 7,9
vWA: 14,19

Culture Medium

IDMEM + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service