Skip to Content
Merck
All Photos(2)

Key Documents

UFSC050SL

Millipore

Amicon® Stirred Cell 50mL Seal Kit (3 each o-ring and gasket)

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031690
NACRES:
NB.24

material

silicone O-ring
silicone gasket

Quality Level

product line

Amicon®

manufacturer/tradename

Amicon®

parameter

50 mL process volume

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

H

11.7 cm (4.6 in.)

filter diam.

44.5 mm

filtration area

13.4 cm2

shipped in

ambient

Principle

Pressure

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Protocols

User guide for Amicon® Stirred cells including features, operation, specifications, and troubleshooting.

User guide for Amicon® Stirred cells including features, operation, specifications, and troubleshooting.

User guide for Amicon® Stirred cells including features, operation, specifications, and troubleshooting.

User guide for Amicon® Stirred cells including features, operation, specifications, and troubleshooting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service