Merck
Aurora B/C-dependent phosphorylation promotes Rec8 cleavage in mammalian oocytes.
Current biology : CB (2022-04-07)
Elvira Nikalayevich, Safia El Jailani, Aude Dupré, Damien Cladière, Yulia Gryaznova, Célia Fosse, Eulalie Buffin, Sandra A Touati, Katja Wassmann
RESUMEN
To generate haploid gametes, cohesin is removed in a stepwise manner from chromosome arms in meiosis I and the centromere region in meiosis II to segregate chromosomes and sister chromatids, respectively. Meiotic cohesin removal requires cleavage of the meiosis-specific kleisin subunit Rec8 by the protease separase.1,2 In yeast and C. elegans, Rec8 on chromosome arms has to be phosphorylated to be cleaved in meiosis I,3-7 whereas Rec8 at the centromere is protected from cleavage by the action of PP2A-B56.8-10 However, in mammalian meiosis, it is unknown whether Rec8 has to be equally phosphorylated for cleavage, and if so, the identity of the relevant kinase(s). This is due to technical challenges, as Rec8 is poorly conserved, preventing a direct translation of the knowledge gained from model systems such as yeast and C. elegans to mammals. Additionally, there is no turnover of Rec8 after cohesion establishment, preventing phosphomutant analysis of functional Rec8. To address the very basic question of whether Rec8 cleavage requires its phosphorylation in mammals, we adapted a biosensor that detects separase activity to study Rec8 cleavage in single mouse oocytes by live imaging. Crucially, through phosphomutant analysis, we identified phosphorylation sites in Rec8 promoting cleavage. We found that Rec8 cleavage depends on Aurora B/C kinase activities and identified an aminoacid residue that is phosphorylated in vivo. Accordingly, inhibition of Aurora B/C kinases during meiotic maturation impairs endogenous Rec8 phosphorylation and chromosome segregation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monofosfato de N6,2′-O-dibutiriladenosina 3′,5′-cíclico sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Anticuerpo anti-fosfo-histona H3 (Ser10), clon 3H10, clone 3H10, Upstate®, from mouse
Sigma-Aldrich
AZD1152-HQPA, ≥98% (HPLC)
Sigma-Aldrich
Anti-GST Tag Antibody, clone DG122-2A7, HRP conjugate, clone DG122-2A7, Upstate®, from mouse