Key Documents
57647
Sample Loop for 9010, 9125, and 9725 Injectors
volume 1 mL, PEEK
About This Item
UNSPSC Code:
41115854
material
PEEK
O.D.
1.6 mm
1/16 in.
volume
1 mL
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)