Saltar a
Z652180
Hettich® MIKRO 200/200R centrifuge accessories
for MIKRO 200/200R and UNIVERSAL 320/320R Centrifuges, Bio-containment lid for rotors 1420, 2424, 2426; 2430
para ver los precios por organización y contrato.
Acerca de este artículo
NACRES:
NB.22
UNSPSC Code:
41103913
Compatibility:
for use with MIKRO 200/200R , for use with UNIVERSAL 320/320R
Manufacturer/tradename:
Hettich 2425
feature
autoclavable aerosol tight
manufacturer/tradename
Hettich 2425
technique(s)
centrifugation: suitable
compatibility
for use with MIKRO 200/200R , for use with UNIVERSAL 320/320R
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Comparar elementos similares
Ver comparación completa
Mostrar Diferencias
1 of 1
Este artículo
Z652199Z652164Z601381
compatibility
for use with MIKRO 200/200R , for use with UNIVERSAL 320/320R
compatibility
for use with MIKRO 200/200R , for use with UNIVERSAL 320/320R
compatibility
-
compatibility
for use with centrifuge models MIKRO 220/220R
manufacturer/tradename
Hettich 2425
manufacturer/tradename
Hettich 2423
manufacturer/tradename
Hettich 2418
manufacturer/tradename
Hettich 1016
technique(s)
centrifugation: suitable
technique(s)
centrifugation: suitable
technique(s)
PCR: suitable, centrifugation: suitable
technique(s)
centrifugation: suitable
feature
autoclavable aerosol tight
feature
-
feature
RCF 12,490, capacity 12,490 mL
feature
capacity 7 ‑ 50 mL, to hold 6 x 50 mL tubes
Still not finding the right product?
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)