Key Documents
Z614734
Vivaspin 20 diafiltration cup
pkg of 12 ea
About This Item
UNSPSC Code:
41104916
NACRES:
NB.24
material
polycarbonate housing
polyethersulfone membrane
sterility
non-sterile
packaging
pkg of 12 ea
manufacturer/tradename
Sartorius VSA005
tube size
~50 mL
working volume
20 mL