Seleccione un Tamaño
| Talla/SKU | Disponibilidad | Precio |
|---|---|---|
5 ea | Comprobar disponibilidad del carrito | $45.400 |
Acerca de este artículo
$45.400
material
polypropylene , yellow
feature
to hold 80 tube(s)
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific HS29025E
L × W × H
225 mm × 67 mm × 28 mm
General description
Features and Benefits
- Easily fill and transfer 80 tubes in compact arrangement (5x16 array)
- Frosted labeling area on bottom of rack
- Embossed Index Numbers for quick sample ID and retrieval
- Made of polypropylene
- Autoclavable
1 of 1
Este artículo | |||
|---|---|---|---|
| description yellow | description green | description natural | description orange rack |
| manufacturer/tradename Heathrow Scientific HS29025E | manufacturer/tradename Heathrow Scientific HS29025C | manufacturer/tradename Heathrow Scientific HS29025A | manufacturer/tradename Heathrow Scientific HS29025F |
| feature to hold 80 tube(s) | feature to hold 80 tube(s) | feature to hold 80 tube(s) | feature to hold 80 tube(s) |
| material polypropylene , yellow | material polypropylene , green | material polypropylene , natural | material orange rack, polypropylene |
| packaging pack of 5 ea | packaging pack of 5 ea | packaging pack of 5 ea | packaging pack of 5 ea |
| L × W × H 225 mm × 67 mm × 28 mm | L × W × H 225 mm × 67 mm × 28 mm | L × W × H 225 mm × 67 mm × 28 mm | L × W × H 226 mm × 67 mm × 28 mm |
Still not finding the right product?
Explore all of our products under Microtube Rack
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?