Documentos clave
EP0030108400
Eppendorf® DNA LoBind Tubes
capacity 0.5 mL, PCR clean, withstand, 30,000 x g, pack of 500 ea
Sinónimos:
Eppendorf® DNA LoBind tubes
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material
cap (push fit)
colorless
conical bottom
description
Low retention
sterility
non-sterile
feature
DNase free
PCR clean
packaging
pack of 500 ea
capacity
0.5 mL
suitability
suitable for PCR
General description
Maximize nucleic acid recovery. Eppendorf LoBind Tubes signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules. The ideal solution for sample preparation and long-term storage of your precious nucleic acids
Features and Benefits
Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)