Documentos clave
1.12937
Supelco
Tap (PE)
for use with 5, 10, 25 L PE canisters with KS 60X6 external thread
About This Item
UNSPSC Code:
41122409
NACRES:
NB.51
compatibility
for use with 5, 10, 25 L PE canisters with KS 60X6 external thread
storage temp.
15-25°C
General description
attachable, self-venting, for 5 l, 10 l and 25 l PE canisters with KS 60x6 external thread