Z693340
Norell® Sample Vault™ NMR tube caps
open port cap, white, diam. 3 mm
About This Item
Código UNSPSC:
41121800
NACRES:
SB.32
Productos recomendados
envase
pkg of 96 ea
fabricante / nombre comercial
Norell SVOP-NORLOC-3-96PK
Diámetro
3 mm
color
white
Categorías relacionadas
Descripción general
Sample Vault caps have superior holding and sealing capabilities which eliminate cap/tube failure in your instrument. The patented design incorporates a white band positioned on the NMR tube that aligns with the base of the cap, indicating a properly locked position. The superior Sample Vault cap attaches semi-permanently for multiple use and for critical applications.
Características y beneficios
- Superior Sample Vault cap attaches semi-permanently for several uses and critical applications.
- Ideal for cold refrigeration storage.
- Suitable for cryo-probes and variable temperature studies.
Información legal
Norell is a registered trademark of Norell, Inc.
Sample Vault is a trademark of Norell, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)