Documentos
Z682500
Ace vapor duct tube, for condenser "V" assembly
joint: ST/NS 29/32, L 45 mm, fits Büchi® models R-114/R-144
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
joint
joint: ST/NS29/32
manufacturer/tradename
Büchi equivalent, 32002
L
45 mm
General description
The vapor duct tube accessories are for Büchi condenser "V" assemblies, and will fit either Büchi Rotavapor model R-210/R-215 or R-114/R-144.
Combi-clips: not included.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG