PL800GS-NIT
Plas-Labs straight sleeves
for 2 piece glove set, fits 8 in. glove box ports, nitrile
Seleccione un Tamaño
$516.800
Fecha estimada de envío02 de abril de 2025
Seleccione un Tamaño
About This Item
$516.800
Fecha estimada de envío02 de abril de 2025
Productos recomendados
material
nitrile
size
8 in.
feature
reusable
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GS-NIT
General description
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
- White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
- Black neoprene rubber for chemical resistance
- Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
- Green nitrile rubber for resistance to oil, fuel, and acids
- Latex rubber for flexibility
Legal Information
For use with
replacement part
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)