Key Documents
GF23296147
Lead-selenium alloy, Pb99Se1
granule, 5mm nominal granule size, alloy pre-cursor, 50g
Sinónimos:
PB106300
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
form
granular
manufacturer/tradename
Goodfellow 232-961-47
weight
50 g
nominal granule size
5 mm