BR140007
BRAND® separating funnel with screw cap
size 1000 mL, polypropylene
Sinónimos:
Funnels, Separating funnels
About This Item
Productos recomendados
material
conical PTFE stopcock
polypropylene
packaging
pack of 1 ea
manufacturer/tradename
BRAND 140007
size
1000 mL
General description
Polypropylene, high clarity for observation of contents. With polypropylene screw cap and special conical PTFE stopcock with retention device. Autoclavable (121 °C).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG