Documentos clave
BR129728
BRAND® PE-LD Media Bottle with screw cap
capacity 50 mL
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material
low-density polyethylene bottle
round bottle
joint
Joints threaded neck (GL32)
feature
graduations: no
with cap
packaging
pack of 100 ea
manufacturer/tradename
BRAND 129728
capacity
50 mL
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)