A100C
Printed Circuit Board Chip
A100C
About This Item
Productos recomendados
material
ENEPIG plating
FR-4 glass epoxy laminate substrate
copper ground plane
copper via(s)
metal pin
plastic header
feature
wirebondable with headers
manufacturer/tradename
A100C