Skip to Content
Merck
All Photos(1)

Key Documents

Z640476

Eppendorf® epT.I.P.S. Reloads, PCR Clean

volume 0.1-10 μL

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
manufacturer/tradename:
Eppendorf® 0030 073.746
Eppendorf® 022491709
L:
34 mm
maximum volume:
10 μL
sterility:
non-sterile
packaging:
pack of 960 (10 trays of 96 tips)

sterility

non-sterile

packaging

pack of 960 (10 trays of 96 tips)

manufacturer/tradename

Eppendorf® 0030 073.746
Eppendorf® 022491709

volume measuring range

0.1-10 μL

technique(s)

PCR: suitable

L

34 mm

maximum volume

10 μL

Looking for similar products? Visit Product Comparison Guide

General description

Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately). PCR clean, free of human DNA, DNase, RNase and PCR inhibitors, batch related certificates available on the internet at http://www.eppendorf.com

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service