Z640433
Eppendorf® epT.I.P.S. Reloads
volume range 50-1000 μL
Synonym(s):
epT.I.P.S. Reload System, pipetting tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
50—1000 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 960 (10 trays of 96 tips)
manufacturer/tradename:
Eppendorf® 0030 073.460
Eppendorf® 022491555
Eppendorf® 022491555
Recommended Products
material
blue
sterility
non-sterile
feature
barrier: no
packaging
pack of 960 (10 trays of 96 tips)
manufacturer/tradename
Eppendorf® 0030 073.460
Eppendorf® 022491555
L
71 mm
maximum volume
1 mL
volume range
50-1000 μL
volume
50—1000 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Eppendorf® epT.I.P.S. Reloads are trays filled with pipette tips designed to refill a reusable box, resulting in reduced waste compared to single-use racks. These refill trays, including the pipette tips, can be autoclaved in the box at temperatures up to 121 °C. Depending on the volume of the pipette tips, the trays in the refill system are either packed against each other or stacked on top of each other. Before picking up the pipette tips, it is necessary to insert the refill trays, including the pipette tips, into the reusable box. Pipettes are available in various packaging units like bulkware, reloads, sterile reloads, racks, or singles.
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately)
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately)
Application
Eppendorf® epT.I.P.S. Reloads are suitable for general laboratory applications to ensure low-contamination transfer of liquids. It has been used for gently mixing the cells.
Features and Benefits
- Single-use pipette tips
- Have a universal cone
- Easy to handle
- Offered in different purity grades
- Color-coded for different volume sizes
- Corresponding color coding to pipettes
- Volume marking enables visual inspection
- Minimal force required for attaching or ejecting
- Provides a secure and complete seal
- Ensures maximum precision and accuracy
- Some volume sizes available with long pipette tips
Other Notes
Eppendorf recommends autoclaving to sterilize the pipette tips.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Measuring key human carbohydrate digestive enzyme activities using high-performance anion-exchange chromatography with pulsed amperometric detection
Barber E, et al.
Nature Protocols (2022)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service