Skip to Content
Merck
All Photos(2)

Key Documents

RAB0310

Sigma-Aldrich

Porcine IL-6 ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

IL-6 ELISA, Porcine IL-6 Detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

porcine

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 45 pg/mL
standard curve range: 40.96-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

Porcine ... IL6(403985)

General description

The Porcine IL-6 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of porcine IL-6 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Porcine IL-6

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rayyan Hemetsberger et al.
Frontiers in cardiovascular medicine, 8, 690476-690476 (2021-07-27)
Objective: We evaluated the short and long-term effect of peri-interventional dabigatran therapy on vasomotion, endothelialization, and neointimal formation in a porcine coronary artery stenting model. Background: Stenting of coronary arteries induces local inflammation, impairs vasomotion and delays endothelialization. Methods: Twenty-eight

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service