Skip to Content
Merck
All Photos(3)

Key Documents

84088

Millipore

Sabouraud Dextrose Agar

NutriSelect® Plus, Glucose 4 %, powder, bottle of 500 g, suitable for microbiology

Synonym(s):

SGA, Sabouraud agar with glucose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
Glucose, 4%
D(+)-glucose, 40 g/L
peptone, 10 g/L

packaging

bottle of 500 g

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

color

light yellow Powder

final pH

5.6±0.2 (25 °C)

application(s)

bioburden testing
cleaning products
clinical testing
cosmetics
environmental
food and beverages
personal care
pharmaceutical

microbiology

suitability

selective for Aspergillus spp.
selective for Candida spp.
selective for Penicillium spp.
selective for Pichia spp.
selective for Saccharomyces spp.
selective for Trichophyton spp.
selective for Zygosaccharomyces spp.
selective for molds (General Media)
selective for yeasts (General Media)

Looking for similar products? Visit Product Comparison Guide

Application

for the cultivation of pathogenic and non-pathogenic fungi, especially dermatophytes.

Preparation Note

Dissolve 65 g in 1 litre distilled water. Autoclave at 121°C for 15 minutes. Do NOT overheat.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald M
Handbook of Media for Clinical and Public Health Microbiology (2013)
Pedro M D S Abrantes et al.
Journal of microbiological methods, 169, 105815-105815 (2019-12-25)
Candida albicans and streptococci are amongst the most common fungal and bacterial organisms present in the oral cavity, with a growing body of evidence implicating C. albicans in increased caries severity and in the formation of the cariogenic biofilm. However
Cathy N John et al.
Frontiers in microbiology, 10, 1021-1021 (2019-06-25)
Background/Objectives: With mucocutaneous candidiasis being highly prevalent in HIV patients, the emergence of fluconazole-resistant Candida species forms a major challenge in treating and eradicating these infections. The objective of this study was to establish the antifungal activity of K21, a

Articles

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service