80521
Trichloroacetyl chloride
for GC derivatization, ≥98.5%
About This Item
Recommended Products
grade
for GC derivatization
Quality Level
vapor pressure
16 mmHg ( 20 °C)
Assay
≥98.5%
98.5-101.5% (AT)
form
liquid
reaction suitability
reagent type: derivatization reagent
reaction type: Acylations
refractive index
n20/D 1.470 (lit.)
n20/D 1.470-1.472
bp
114-116 °C (lit.)
mp
−57 °C (lit.)
density
1.629 g/mL at 25 °C (lit.)
SMILES string
ClC(=O)C(Cl)(Cl)Cl
InChI
1S/C2Cl4O/c3-1(7)2(4,5)6
InChI key
PVFOMCVHYWHZJE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
213.8 °F - open cup
Flash Point(C)
101 °C - open cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service