Merck
Trichloroacetylation of some cyclic enamines.
Chemical & pharmaceutical bulletin (2003-04-04)
Hülya Senöz, Nazan Tunoğlu
ABSTRACT
The pyrrolidine and morpholine enamines of cyclic ketones such as cyclohexanone and cyclopentanone were successfully diacetylated at alpha- and alpha'-positions with trichloroacetyl chloride using zinc catalyst. Morpholine enamines of the cyclic ketones gave acetylated morpholine in good yields besides the corresponding diacetylated cyclic enamines. When the same reactions were performed by using triethylamine without using zinc, monoacetylation products of the same enamines were synthesized.
Sigma-Aldrich
Trichloroacetyl chloride, 99%
Supelco
Trichloroacetyl chloride, for GC derivatization, ≥98.5%