Skip to Content
Merck
All Photos(2)

Key Documents

EZFITSAM1

Millipore

EZ-Fit® Manifold

1-place manifold, for use with EZ-Fit® Filtration Unit, 316L stainless steel

Synonym(s):

EZ-Fit® Filtration Manifold, EZ-Fit® manifold for EZ-Stream® vacuum pump, high throughput application, Filtration Manifold Base, Manifold Base, Manifold head

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

Quality Level

material

316L stainless steel

Sterilization Compatibility

autoclavable compatible

feature

1-place manifold

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® Filtration Unit

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

EZ-Fit® Filtration Manifold Base with 1 place is uniquely designed to carry out laboratory filtration of samples. The filtration manifold construction eliminates all sources of contamination over repeated use as each part of the manifold base is cleanable and autoclavable.
The EZ-Fit® Filtration Manifold Base can accommodate one filtration head and the ergonomic design enables operation under laminar flow hood and save laboratory bench space.
The 2-way valve system of the filtration manifold prevents the user to accidentally touch the filtration head while opening or closing the valve.

Application

EZ-Fit® Filtration Manifold Base with 1 place is used to carry out routine laboratory filtration samples. Liquid samples of raw materials, in process and finished products from diverse industries can be tested using the equipment.

Features and Benefits

The following features and benefits are associated with EZ-Fit® Filtration Manifold Base:
  • Different filtration heads fit both reusable and disposable filtration devices.
  • Each component is removable for deeper cleaning.
  • Autoclavable parts.
  • Quick fit connections for vacuum tubing.
  • Low height for easier use inside Laminar Flow Hood.

Packaging

EZFITBASE1 x1
EZFITMHTA1 x1

Analysis Note

The EZ-Fit® Filtration Manifold Base with 1 place can filter 1 test sample per cycle.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

configured for

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The EZ Product Family optimizes your microbial testing workflow to deliver reliable and reproducible microbial enumeration test results. The product lineup consists of filtration devices, manifolds, calibrated vacuum sources, and rapid microbial detection platforms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service