EFHAW25BS
EZ-Fit® filtration unit without absorbent pad
pore size 0.45 μm, mixed cellulose esters (MCE) membrane, white filter, volume 250 mL, pkg of Bulk packaging with protective bag, suitable for (suitable for solid cultivation)
Synonym(s):
Membrane filtration device made of gridded, mixed cellulose esters membrane for bioburden testing
About This Item
Recommended Products
material
gridded filter
mixed cellulose esters (MCE) membrane
styrene-butadiene device
white filter
Quality Level
Agency
ISO 7004
sterility
electron beam irradiated
feature
ready-to-use
manufacturer/tradename
EZ-Fit®
packaging
pkg of Bulk packaging with protective bag
capacity
250 mL
filter diam.
47 mm
funnel capacity
250 mL
volume
250 mL
pore size
0.45 μm pore size
input
sample type food(s)
sample(s) (environmental)
sample type pharmaceutical(s)
sample type beverage(s)
suitability
suitable for (suitable for solid cultivation)
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
compatibility
for use with EZ-Fit® System
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
The filtration units are e-beam irrated, a vented lid covers the sample throughout filtration, and a 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer, to protect samples from contamination while ensuring reliable microbial enumeration results.
The EZ-FIT® Azure feature makes the EZ-FIT® filtration unit even more ergonomic and convenient with transparent funnel and all-around filling level indicators for an easy filling from any angle. The innovative funnel assembly design prevents any leaks or sample by-pass of the membrane.
The filtration units conform to international standards and minimize sample residue. The units can be stacked to save the work area. This disposable device is designed to fit onto the EZ-FIT® Manifold, which features specific EZ-FIT® Filtration heads.
Application
Legal Information
configured for
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.
The EZ Product Family optimizes your microbial testing workflow to deliver reliable and reproducible microbial enumeration test results. The product lineup consists of filtration devices, manifolds, calibrated vacuum sources, and rapid microbial detection platforms.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service