Skip to Content
Merck
All Photos(1)

Key Documents

688002-M

Millipore

Y-27632

≥95% (HPLC), liquid, Rho kinase inhibitor, Calbiochem®

Synonym(s):

InSolution Y-27632 in DMSO, Rho Kinase Inhibitor VI, (R)-(+)- trans-N-(4-Pyridyl)-4-(1-aminoethyl)-cyclohexanecarboxamide, 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N3O · 2HCl · xH2O
Molecular Weight:
320.26 (anhydrous basis)
UNSPSC Code:
12352200

Product Name

Y-27632 in DMSO, InSolution, ≥95%, 10 mM, reversible and selective inhibitor of Rho-associated protein kinases

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

General description

A highly potent, cell-permeable, reversible, and selective inhibitor of Rho-associated protein kinases (Ki = 140 nM for p160ROCK). Also inhibits ROCK-II with equal potency. The inhibition is competitive with respect to ATP. Exhibits 10- to 50-fold lower affinity for PKCε than p160ROCK. Does not affect the activity of p21-activated protein kinase (PAK) even at higher concentrations (~100 µM). Also acts as a potent inhibitor of agonist-induced Ca2+ sensitization of myosin phosphorylation and smooth muscle contraction. Prevents apoptosis and enhances the survival and cloning efficiency of dissociated hES cells without affecting their pluripotency.

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg/296 µl) solution of Y-27632 (Cat. No. 688000)in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Watanabe, K., et al. 2007. Nature Biotech.25, 681.
Chitaley, K., et al. 2001. Nat. Med.7, 119.
Davies, S.P., et al. 2000. Biochem. J. 351, 95.
Narumiya, S., et al. 2000. Methods Enzymol.325, 273.
Hirose, M., et al. 1998. J. Cell Biol.141, 1625.
Maekawa, M., et al. 1999. Science285, 895.
Uehata, M., et al. 1997. Nature389, 990.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service