Skip to Content
Merck
All Photos(1)

Key Documents

681669

Sigma-Aldrich

IWR-1-endo

≥95% (HPLC), solid, Wnt antagonist, Calbiochem®

Synonym(s):

Wnt Antagonist I, IWR-1-endo, Inhibitor of Wnt Response-1, Wnt Pathway Inhibitor I, TNKS1/2 Inhibitor II, Tankyrase1/2 Inhibitor II, Wnt Pathway Inhibitor I, Inhibitor of Wnt Response-1, Tankyrase1/2 Inhibitor II, TNKS1/2 Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19N3O3
Molecular Weight:
409.44
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Wnt Antagonist I, IWR-1-endo, Wnt Antagonist I, IWR-1-endo, is a cell-permeable inhibitor of TNKS1/PARP5a (IC₅₀ = 131 nM) & TNKS2/PARP5b (IC₅₀= 56 nM). Suppress Wnt-stimulated transcription activity (IC₅₀ = 180 nM).

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable p-imidobenzamidoquinoline, endo-diastereomer that is shown to inhibit the activity of TNKS1/PARP5a and TNKS2/PARP5b in in vitro auto-PARsylation assays (IC50 = 131 and 56 nM, respectively) and effectively suppress Wnt-stimulated transcription activity in L-Wnt-STF-based reporter assays (IC50 = 180 nM), while exhibiting little activity against PARP1 or PARP2 (IC50 >18.75 µM). Although both IWR-1-endo and XAV939 act as reversible Wnt pathway inhibitors and exhibit similar pharmacological effects both in vitro and in vivo, IWR-1-endo exerts its effect via interaction with Axin, while XAV939 binds TNKS directly.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chen, B., et al. 2009. Nature Chem. Biol.5, 100.
Huang, S.M., et al. 2009. Nature461, 614.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanping Liu et al.
Aging, 12(8), 7411-7430 (2020-04-29)
Human pluripotent stem cell-derived cardiomyocytes (hPSC-CMs) have great potential in biomedical applications. However, the immature state of cardiomyocytes obtained using existing protocols limits the application of hPSC-CMs. Unlike adult cardiac myocytes, hPSC-CMs generate ATP through an immature metabolic pathway-aerobic glycolysis
Mei-Ling Gao et al.
Frontiers in cell and developmental biology, 8, 128-128 (2020-03-27)
Although an increasing number of disease genes have been identified, the exact cellular mechanisms of retinitis pigmentosa (RP) remain largely unclear. Retinal organoids (ROs) derived from the induced pluripotent stem cells (iPSCs) of patients provide a potential but unvalidated platform
Mingzheng Wu et al.
Pain, 161(3), 532-544 (2019-11-19)
Preventing and treating opioid dependence and withdrawal is a major clinical challenge, and the underlying mechanisms of opioid dependence and withdrawal remain elusive. We hypothesized that prolonged morphine exposure or chronic inflammation-induced μ-opioid receptor activity serves as a severe stress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service