Skip to Content
Merck
All Photos(1)

Key Documents

5.05881

Sigma-Aldrich

(R)-(+)-SKF38393 Hydrochloride

Synonym(s):

(R)-(+)-SKF38393 Hydrochloride, Dopamine D₁/D₅ Receptor Partial Agonist, (R)-SKF-38393 Hydrochloride, SKF-38393, SKF38393, SKF 38393

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO2 · xHCl
CAS Number:
Molecular Weight:
255.31 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 100 mM

storage temp.

−20°C

SMILES string

Cl.N1C[C@@H](c3c(cc(c(c3)O)O)CC1)c2ccccc2

InChI

1S/C16H17NO2.ClH/c18-15-8-12-6-7-17-10-14(13(12)9-16(15)19)11-4-2-1-3-5-11;/h1-5,8-9,14,17-19H,6-7,10H2;1H/t14-;/m1./s1

InChI key

YEWHJCLOUYPAOH-PFEQFJNWSA-N

General description

A benzazepine derivative that acts as a selective Dopamine D1/D5 receptor partial agonist. Examination of the isomers in several in vitro and in vivo tests for both central and peripheral dopaminergic activity revealed that activity resided almost exclusively in the R isomer. The results suggest that the properly oriented 1-phenyl substituent of 1 is important for dopamine-like activity. Reported to have stimulant and anorectic effects.

Biochem/physiol Actions

Primary Target
D₁

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Trampus, M. et al. 1993. J. Pharmacol.235, 83.
Cooper, S. J. et al. 1990. Psychopharmacol.100, 182.
Dubois, A. et al. 1986. Neurosci.19, 125.
Kaiser, C. C. et al. 1982. J. Med. Chem.25, 697.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service