692018
3-Methylphenethyl bromide
96%
Synonym(s):
1-(2-Bromoethyl)-3-methylbenzene, 1-Bromo-2-m-tolylethane
About This Item
Recommended Products
Assay
96%
form
liquid
refractive index
n20/D 1.550
bp
101-103 °C/11 mmHg
density
1.307 g/mL at 25 °C
functional group
bromo
SMILES string
Cc1cccc(CCBr)c1
InChI
1S/C9H11Br/c1-8-3-2-4-9(7-8)5-6-10/h2-4,7H,5-6H2,1H3
InChI key
STQJFINZGUXVEX-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
228.9 °F
Flash Point(C)
109.4 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service