Skip to Content
Merck
All Photos(6)

Key Documents

HPA003898

Sigma-Aldrich

Anti-MCM7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CDC47, Anti-MCM2, Anti-PPP1R104

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

QSPTFMPLIMCPSQECQTNRSGGRLYLQTRGSRFIKFQEMKMQEHSDQVPVGNIPRSITVLVEGENTRIAQPGDHVSVTGIFLPILRTGFRQVVQGLLSETYLEAHRIVKMNKSEDDESGAGE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCM7(4176)

Immunogen

DNA replication licensing factor MCM7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DNA replication licensing factor MCM7 is a protein encoded by the MCM7 gene in humans. MCM7 (mini-chromosome maintenance protein 7) is very essential for the initiation of genomic replication as a component of the pre-replication complex. It might play an oncogenic role in gastric tumorigenesis and serves as a potential prognostic marker. It can be a potential prognostic marker and a novel therapeutic target in glioblastoma multiforme (GBM) therapy as well. Its expression may be a useful predictor of prognosis in HCC (hepatocellular carcinoma) patients after resection.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86671

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan-Ming Zhou et al.
Liver international : official journal of the International Association for the Study of the Liver, 32(10), 1505-1509 (2012-07-13)
Dysregulation of minichromosome maintenance protein 7 (MCM7) was previously identified in multiple human malignancies. The clinical significance of MCM7 expression is yet to be delineated in patients with hepatocellular carcinoma (HCC). Paired cancerous and non-cancerous specimens from 87 patients with
E P Erkan et al.
Oncogene, 33(39), 4778-4785 (2013-10-30)
Minichromosome maintenance (MCM) proteins are key elements that function as a part of the pre-replication complex to initiate DNA replication in eukaryotes. Consistent with their roles in initiating DNA replication, overexpression of MCM family members has been observed in several
Wei Kang et al.
Oncology reports, 31(5), 2071-2078 (2014-03-22)
MCM7 (mini-chromosome maintenance protein 7) is essential for the initiation of genomic replication as a component of the pre-replication complex. The present study aimed to analyze its expression, clinical significance and biological functions in gastric adenocarcinoma (GAC). The MCM7 protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service