Key Documents
AXYSCT200SSYS
Self standing screw cap tubes and attached caps with O-rings
capacity 2.0 mL, yellow, sterile
Synonym(s):
centrifuge tubes
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
screw top cap
yellow
sterility
sterile
feature
RCF 20,000 × g
packaging
pack of 500 tubes
case of 8 packs
manufacturer/tradename
Axygen Inc
Corning
capacity
2.0 mL