FA10006M
PIPETMAN P1000L Micropipette Metal eject
for use with BioControl® GDS, suitable for PCR, tip volume 100-1000 μL
Synonym(s):
Micropipette
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121521
NACRES:
NB.01
packaging
box of 1 unit
technique(s)
PCR: suitable
tip volume
100-1000 μL
compatibility
for use with BioControl® GDS
storage temp.
room temp
General description
PIPETMAN® Micropipette with metal eject is an air-cushion pipette with outstanding precision and accuracy. It is extremely reliable and robust, allowing for long service life. The PIPETMAN® provides fast and ergonomic handling with reduced operating efforts and active aerosol reduction. As part of the manual sample preparation for Assurance® GDS assays, a single channel pipette with 100-1000 μL pipetting capacity is comfortable to use and provides ideal functionality to complete reagent and sample preparation for GDS PCR. The metal ejector enables easy disposal of the tips with minimal force.
Application
PIPETMAN® Micropipette with metal eject is used with Assurance® GDS to complete sample preparation for the detection of pathogenic organisms in foods, ingredients, and environmental samples.
Components
The Product Contains the following:
- Single Micropipette.
- Metal Ejector.
Legal Information
Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
BioControl is a registered trademark of BioControl Systems, Inc.
Pipetman is a registered trademark of Gilson S.A.S. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service