Skip to Content
Merck
All Photos(1)

Documents

Y0000450

Ketamine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Ketamine hydrochloride, (±)-2-(2-Chlorophenyl)-2-(methylamino)cyclohexanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16ClNO · HCl
CAS Number:
Molecular Weight:
274.19
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ketamine

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CNC1(CCCCC1=O)c2ccccc2Cl

InChI

1S/C13H16ClNO.ClH/c1-15-13(9-5-4-8-12(13)16)10-6-2-3-7-11(10)14;/h2-3,6-7,15H,4-5,8-9H2,1H3;1H

InChI key

VCMGMSHEPQENPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ketamine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulo R Shiroma et al.
The international journal of neuropsychopharmacology, 17(11), 1805-1813 (2014-06-26)
The N-methyl-D-aspartate glutamate receptor antagonist ketamine has demonstrated rapid antidepressant effects in treatment-resistant depression (TRD). However, evaluation of ketamine's neurocognitive aspects in TRD has started to be explored. This study aims to (1) examine baseline neurocognitive performance and change in
Jana Sawynok
Anesthesia and analgesia, 119(1), 170-178 (2014-06-20)
Ketamine, in subanesthetic doses, produces systemic analgesia in chronic pain settings, an action largely attributed to block of N-methyl-D-aspartate receptors in the spinal cord and inhibition of central sensitization processes. N-methyl-D-aspartate receptors also are located peripherally on sensory afferent nerve
Yongzhi Wang et al.
Shock (Augusta, Ga.), 42(4), 343-349 (2014-07-01)
Systemic inflammatory response syndrome and severe infections are associated with major derangements in the coagulation system. The purpose of this study was to examine the dynamic alterations in thrombin generation in abdominal sepsis. Abdominal sepsis was induced by cecal ligation
José Luis Calvo-Guirado et al.
Clinical oral investigations, 18(5), 1495-1505 (2013-10-19)
This study presents a biomechanical comparison of bone response to commercially pure titanium screws with four different types of surface topographies placed in the tibial metaphysis of 30 rabbits. One hundred twenty implants were tested double-blinded: (a) blasted, acid-etched, and
Yongzhi Wang et al.
American journal of physiology. Heart and circulatory physiology, 308(5), H540-H547 (2014-12-17)
Abdominal sepsis is associated with significant changes in systemic inflammation and coagulation. The purpose of the present study was to examine the role of peripheral blood monocytes for systemic coagulation, including thrombin generation and consumption of coagulation factors. Abdominal sepsis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service