CDFC01204
Super-C Carbon Cartridge
For use with Milli-Q® and Milli-RO™ Bowl systems
Synonym(s):
Super-C Carbon Cartridge 30 cm (12")
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
feature
removes organics from water pretreated by reverse osmosis, deionization or distillation
packaging
pkg of 4 units
L
30 cm (12 in.)
total organic carbon (TOC) residue
<20 ppb
compatibility
for use with Alpha-Q
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Super-C Carbon Cartridge 30 cm (12") is a replacement cartridge used to remove organics from water pre-treated by reverse osmosis (RO), deionization or distillation. Use in combination with Rogard II prefilter, CDPRM1206.
For use with Milli-Q® and Milli-RO™ bowl-style systems.
For use with Milli-Q® and Milli-RO™ bowl-style systems.
Features and Benefits
Purification method involves the use of activated carbon-based adsorbent.
Legal Information
MILLI-RO is a trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service