Skip to Content
Merck
All Photos(1)

Key Documents

1.00214

Millipore

GranuCult® prime Columbia Agar (base)

according to ISO 10272, according to EP, according to USP, according to JP, suitable for microbiology

Synonym(s):

Columbia Agar (CA), Columbia Agar Base (CAB), Columbia Blood Agar (CBA)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

according to EP
according to ISO 10272
according to JP
according to USP

Quality Level

Sterilization Compatibility

autoclavable compatible

form

powder

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® prime

storage condition

protect from light (Keep dry and tightly closed)

technique(s)

microbe id | specific enzyme detection: suitable
microbiological culture: suitable

color

beige

pH

7.3±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

microbiology

storage temp.

15-25°C

suitability

Enterococcus spp.
Haemophilus spp.
Pneumococcus spp.
Staphylococcus spp.
Streptococcus spp.

General description

GranuCult® prime Columbia Agar (base) can be used with or without supplements as a highly nutritious general-purpose medium. It provides superior growth-supporting properties due to its mixture of peptones, including pancreatic digest of casein, peptic digest of meat, and pancreatic digest of the heart. Yeast extract and maize starch support as energy and vitamin sources. Agar is the solidifying agent.When supplemented with blood, e.g., sheep, rabbit, or horse blood, microorganisms grow on this medium showing their typical hemolytic reactions. Fresh, defibrinated sheep blood is most suitable for determining the different hemolysis forms. Heated blood agar (‘chocolate agar’) is an extremely rich culture medium and can be prepared by heating the medium after the blood has been added.Columbia agar base can be used to prepare Lactose milk agar, and egg-yolk agar [Lactose egg yolk (LEY) agar, Willis and Hobbs agar] for isolating and differentiating the strains of Clostridium spp. according to their ability to produce lecithinase and/or lipase, to ferment the carbohydrate lactose.

Application

GranuCult® prime Columbia Agar (base) is suitable for the isolation and cultivation of fastidious microorganisms from the food chain, water, pharmaceutical, and other materials; and for the determination of hemolytic reactions after the addition of defibrinated blood.

Features and Benefits

  • GranuCult® offers superior granulated culture media.
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment.
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions.
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Storage and Stability

  • Acc. to ISO 17995, a self-prepared base medium (without added blood) can be stored in airtight bottles in the dark and protected against evaporation at (5 ± 3 °C) for up to one month.
  • Acc. to ISO 10272-1/-2 and ISO 17995, self-prepared plates with the addition of blood can be stored in the dark and protected against evaporation at (5 ± 3 °C) for up to one month.

Other Notes

  • Do not use clumped or discolored medium.
  • The prepared medium without the addition of supplements is clear to slightly opalescent and yellowish-brown.
  • Before inoculation, allow the prepared medium to equilibrate at room temperature if it is  stored at a lower temperature. 
  • There should be no visible moisture on the plates before use. When moisture is present, the plates should be dried for the minimum time required to remove visible moisture, following the procedure as described by EN ISO 11133.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service