1.03784
Egg yolk emulsion
pasteurized, 50%, suitable for microbiology
Synonym(s):
Egg yolk suspension, EY broth, egg yolk enrichment broth
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106212
NACRES:
NA.74
Recommended Products
biological source
chicken egg yolk
Quality Level
form
emulsion
packaging
pkg of 10 × 100 mL
concentration
50%
technique(s)
microbiological culture: suitable
density
1.02 g/cm3
application(s)
microbiology
storage temp.
2-8°C
General description
Egg yolk emulsion is a mixture of egg yolk, typically from chicken eggs, and an aqueous solution, usually saline or phosphate-buffered saline (PBS). It is commonly used in microbiology and related fields as a growth supplement for bacteria and other microorganisms. The emulsion provides a complex mixture of nutrients, including lipids, amino acids, and vitamins, that support the growth of a wide range of microorganisms. Egg yolk emulsion can be added to a variety of microbial culture media to improve the growth of fastidious bacteria that require specific nutrients or additional growth factors. It is particularly useful for the cultivation of bacteria that need cholesterol for growth, such as Mycobacterium species and some of the anaerobic bacteria.
Application
Egg yolk emulsion is commonly used for the cultivation of Mycobacterium tuberculosis and other Mycobacterium species, as well as for the growth of anaerobic bacteria. In addition, egg yolk emulsion can be used as a component of enrichment broths to improve the recovery of pathogens from food samples. Egg yolk emulsion can also be used as an ingredient in selective media formulations, such as egg yolk-polymyxin agar, used for the selective isolation of anaerobic bacteria from various samples.
Linkage
Replaces: 1.03784.0007
Analysis Note
Sterility test: sterile
Typical composition: Sterile egg-yolk 500 ml; NaCl 4.25 g; distilles water to give a final volume of 1000 ml.
Growth promotion test in accordance with ISO 11133.
Egg-yolk Emulsion tested in Mannitol-Egg-yolk-Polymyxine -Agar (=MYP-agar) Art. 1.05267.
Growth
- Bacillus cereus ATCC 11778 (WDCM 00001):
good growth
- Bacillus subtilis ATCC 6633 (WDCM 00003): no limit
- Escherichia coli ATCC 8739 (WDCM 00012): no growth
- Escherichia coli ATCC 25922 (WDCM 00013): no growth
Colony colour
- Bacillus cereus ATCC 11778 (WDCM 00001):
pink
- Bacillus subtilis ATCC 6633 (WDCM 00003): yellow
Precipitate
- Bacillus subtilis ATCC 6633 (WDCM 00003):
-
- Bacillus cereus ATCC 11778 (WDCM 00001): +
Incubation: 48 hrs. 30 °C; B.cereus 24-48 hrs. 30 °C.
Typical composition: Sterile egg-yolk 500 ml; NaCl 4.25 g; distilles water to give a final volume of 1000 ml.
Growth promotion test in accordance with ISO 11133.
Egg-yolk Emulsion tested in Mannitol-Egg-yolk-Polymyxine -Agar (=MYP-agar) Art. 1.05267.
Growth
- Bacillus cereus ATCC 11778 (WDCM 00001):
good growth
- Bacillus subtilis ATCC 6633 (WDCM 00003): no limit
- Escherichia coli ATCC 8739 (WDCM 00012): no growth
- Escherichia coli ATCC 25922 (WDCM 00013): no growth
Colony colour
- Bacillus cereus ATCC 11778 (WDCM 00001):
pink
- Bacillus subtilis ATCC 6633 (WDCM 00003): yellow
Precipitate
- Bacillus subtilis ATCC 6633 (WDCM 00003):
-
- Bacillus cereus ATCC 11778 (WDCM 00001): +
Incubation: 48 hrs. 30 °C; B.cereus 24-48 hrs. 30 °C.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service