Skip to Content
Merck
All Photos(1)

Key Documents

W6624

Pyruvic -2-13C acid

API for Clinical Studies, ≥99 atom % 13C, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH313COCO2H
CAS Number:
Molecular Weight:
89.05
MDL number:
UNSPSC Code:
12352115


Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

grade

cGMP

isotopic purity

≥99 atom % 13C

Assay

≥95% (CP)

form

viscous liquid

bp

165 °C

mp

11-12 °C

mass shift

M+1

shipped in

dry ice

storage temp.

-10 to -25°C

SMILES string

C[13C](=O)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Packaging

This product is available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian T Chung et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 309, 106617-106617 (2019-10-28)
We developed methods for the preparation of hyperpolarized (HP) sterile [2-13C]pyruvate to test its feasibility in first-ever human NMR studies following FDA-IND & IRB approval. Spectral results using this MR stable-isotope imaging approach demonstrated the feasibility of investigating human cerebral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service