Merck
GC Analysis of Free Fatty Acids in Parmesan Cheese using Nukol after SPME using 65 μm Carbowax/DVB Fiber
GC Analysis of Free Fatty Acids in Parmesan Cheese using Nukol after SPME using 65 μm Carbowax/DVB Fiber suitable for GC, application for SPME
Materiali
Colonna analitica
N° Catalogo
Descrizione
Determinazione del prezzo
Nukol Capillary GC Column
size × I.D. 15 m × 0.25 mm, df 0.25 μm
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Supporto per fibra SPME
for use with Manual Sampling, pk of 1
Supporto per fibra SPME
for use with CTC CombiPAL, Gerstel MPS2 Autosamplers, pk of 1
Standard
N° Catalogo
Descrizione
Determinazione del prezzo
Valeric acid
analytical standard
Butyric acid
analytical standard
Decanoic acid
analytical standard
Hexanoic acid
analytical standard
Nonanoic acid
analytical standard
Octanoic acid
analytical standard
Propionic acid
analytical standard
Dodecanoic acid
analytical standard
Isobutyric acid
analytical standard
Heptanoic acid
analytical standard
Isovaleric acid
analytical standard
CONDITIONS
sample/matrix
100 mg cheese in 40 mL vial
SPME fiber
65 μm Carbowax/divinylbenzene StableFlex (57336-U)
extraction
65 °C 15 min (headspace)
desorption process
250 °C, 1 min
column
Nukol, 15 m × 0.25 mm, 0.25 μm (24106-U)
oven
50 °C (2 min) to 220 °C at 10 °C/min
inj. temp.
250 °C
injection
splitless/split (closed 1 min), 250°C
detector
FID, 260 °C
carrier gas
helium, 30 cm/sec
liner
0.75 mm liner
sample
Parmesan cheese
Descrizione
Risultati analitici
In this study, SPME was used for the analysis of free fatty acids in Parmesan cheese using a 65 μm Carbowax/divinylbenzene (DVB) SPME fiber. Headspace extraction of the cheese sample was conducted at 65 °C for 15 minutes and analyzed by GC with FID detection. SPME is ideal for analyzing the volatiles associated with solid food samples. The phase chemistry of the Nukol GC column provides excellent peak shape of acidic compounds.