Merck
GC Analysis of Volatiles in Water on Equity-1 after SPME using 30 μm PDMS Fiber
GC Analysis of Volatiles in Water on Equity-1 after SPME using 30 μm PDMS Fiber suitable for GC, application for SPME
Materiali
Fibra per SPME
N° Catalogo
Descrizione
Determinazione del prezzo
Unità fibra SPME polidimetilsilossano (PDMS)
df 30 μm(PDMS, needle size 24 ga, for use with autosampler
CONDITIONS
sample/matrix
0.7 mL water spiked with 2-600 μg/liter each analyte, 0.25 g NaCl added
SPME fiber
30 μm PDMS (57309)
extraction
headspace, 12 sec (no stirring), ambient temp.
desorption process
2-3 min, 250 °C (0.75-1.0 mm I.D. inj. port liner)
column
Equity-1, 10 m × 0.20 mm I.D., 1.2 μm (available on request)
oven
70 °C (0.2 min) to 180 °C at 50 °C/min
detector
FID, 250 °C