Column Coupler 55213 can be used with Supelco brand columns SUPELCOSIL, Discovery, Discovery HS, Discovery Bio Wide Pore, Ascentis, Astec Cyclobond, Astec Chirobiotic, and Hamilton analytical size columns. Remove the outlet fitting of the first column in line, the inlet fitting of the second column in line, and then couple the two columns together.
55213
HPLC Column Coupler
for use with Discovery and SUPELCOSIL Columns, 3.0, 4.0, 4.6 mm I.D.
About This Item
Prodotti consigliati
Nome del prodotto
HPLC Column Coupler,
Compatibilità
for use with Discovery and SUPELCOSIL columns, 3.0, 4.0, 4.6 mm I.D.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
-
What does product 55213 look like and which columns is it used to connect?
1 answer-
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.