Documenti fondamentali
509892
SGE Microvolume Syringe for Rheodyne® and Valco® Valves, Removable Needle
100R-GT-LC, volume 100 μL, needle L × O.D. 2 in. × 0.7112 mm (0.028 in.)
About This Item
Codice UNSPSC:
41122001
Materiali
blunt tip stainless steel needle (point style 3)
Caratteristiche
Removable needle
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
SGE 005312
Lungh. × D.E. ago
2 in. × 0.7112 mm (0.028 in.)
Volume
100 μL
Compatibilità
for use with SGE RN
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note legali
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Valco is a registered trademark of Valco Instruments Co. Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)