Documenti fondamentali
502413
Upchurch PEEK Microtight® Connector
Butt connects 2 pieces of capillary. True zero dead volume design, pkg of 2 ea
Sinonimo/i:
Microtight® Union
About This Item
Codice UNSPSC:
41115700
Descrizione
zero dead volume
Confezionamento
pkg of 2 ea
Descrizione generale
Connects 2 pieces of capillary tubing. True zero-dead volume design -- tubing pieces butt together. Use gauge plug (included) to properly seat tubing. Includes 2 MicroFingertight fittings.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note legali
Microtight is a registered trademark of IDEX Health & Science, LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)