Documenti
23779
Supelco
Capillary Column Tags
pkg of 100 ea
About This Item
Codice UNSPSC:
41115700
Materiali
0.016″ aluminum
Confezionamento
pkg of 100 ea
Lungh. linguetta
5 1/4 in.
Lungh. × largh. superficie utile
1 1/8 in. × 3/4 in.
Descrizione generale
Permanently label capillary columns.
  • Soft enough to imprint with a ball-point pen
  • Big enough for all necessary information
  • Lightweight and will not damage column
Choose from one of the most recent versions:
Certificati d'analisi (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Servizio Clienti