Documenti fondamentali
Z764663
MAGic Clamp bottle rack
for 4 x 500 or 600 mL bottles (max. 4 tubes), pkg of 1 ea
Sinonimo/i:
bottle clamps
Scegli un formato
1 EA
CHF 233.00
CHF 233.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 233.00
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.77
CHF 233.00
Check Cart for Availability
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Benchmark Scientific H1000-MR-T600
Descrizione generale
The convenient MAGic Clamp platforms (large or small) are available for use with Erlenmeyer flasks and test tube racks. The unique magnetic attachment method is widely recognized as the easiest way to instantly change between flask clamps of different sizes.
Note legali
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)