Documenti fondamentali
Z763411
MAGic Clamp universal platform for flasks and tube racks
L × W 14 in. × 12 in.
Sinonimo/i:
magnetic clamps, orbital shaker platforms
Scegli un formato
1 EA
CHF 356.00
CHF 356.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 356.00
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.77
CHF 356.00
Check Cart for Availability
Produttore/marchio commerciale
Benchmark Scientific BT3000-MR
Lungh. × largh.
14 in. × 12 in.
Descrizione generale
The convenient MAGic Clamp platforms (large or small) are available for use with Erlenmeyer flasks and test tube racks. The unique magnetic attachment method is widely recognized as the easiest way to instantly change between flask clamps of different sizes.
Note legali
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)