Documenti fondamentali
Z757772
Sartorius filter pipette tips
volume range 0.5-20 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Sinonimo/i:
filter pipette tips, pipetting tips, pipettor tips
Scegli un formato
960 EA
CHF 191.00
CHF 191.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
960 EA
CHF 191.00
About This Item
Codice UNSPSC:
41121601
NACRES:
NB.01
CHF 191.00
Check Cart for Availability
Materiali
colorless
polypropylene (Virgin)
Sterilità
sterile
Caratteristiche
DNase free
RNase free
barrier
filter
Confezionamento
pack of 960 ea (10 racks of 96 tips)
Produttore/marchio commerciale
Sartorius 790021F
Volume massimo
20 μL
Intervallo di volume
0.5-20 μL
Volume
0.5—20 μL
Tipo di binding
low binding surface
Descrizione generale
The Sartorius filter tips of the Biohitfamily are made of polypropylene and feature a polyethylene filter barrier to prevent aerosol and liquid contamination. This filter helps protect against the rist of cross-contamination and reduces maintenance requirements of the pipette. The filter tips are suited for molecular biology, microbiology, cell culture and radioactive work.
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
  • Pipette foaming liquids like buffers and proteins
  • Pipette solvents
  • Reverse pipetting
  • Multiple dispensing functions of electronic pipettes
Altre note
Browse all pipette tips
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)